accessibility menu, dialog, popup

UserName

CAS RN 4421-08-3

H3COHON

CAS RN 4421-08-3

IUPAC Name: 4-hydroxy-3-methoxybenzonitrile
Synonyms: 4-hydroxy-3-methoxybenzonitrile
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: QJRWLNLUIAJTAD-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1O)C#N
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)