missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxybenzonitrile, 98%, ACROS Organics™

Product Code. 10042190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10042190 25g; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10042190 Supplier Thermo Scientific Acros Supplier No. 172030250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4421-08-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 85°C to 89°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C8H7NO2
Linear Formula HOC6H3(OCH3)CN
MDL Number MFCD00001820
Quantity 25 g
Beilstein 10, 398
Synonym vanillonitrile, m-anisonitrile, 4-hydroxy, 4-hydroxy-3-methoxy-benzonitrile, 3-methoxy-4-hydroxybenzonitrile, 4-hydroxy-3-methoxybenzenecarbonitrile, pubchem13112, 2-methoxy-4-cyanophenol, 4-cyano-2-methoxyphenol, 5-cyano-2-hydroxyanisole, 2-methoxy-4-cyano-phenol
InChI Key QJRWLNLUIAJTAD-UHFFFAOYSA-N
SMILES COC1=C(C=CC(=C1)C#N)O
IUPAC Name 4-hydroxy-3-methoxybenzonitrile
Molecular Weight (g/mol) 149.15
PubChem CID 78135
Formula Weight 149.15
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-methoxybenzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.