accessibility menu, dialog, popup

UserName

H3COFOHF

CAS RN 443-42-5

IUPAC Name: 3,5-difluoro-4-methoxyphenol
Synonyms: 3,5-difluoro-4-methoxyphenol
Molecular Weight (g/mol): 160.12
Molecular Formula: C7H6F2O2
InChi Key: WPNHIYWMMCVBGE-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(O)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results