accessibility menu, dialog, popup

UserName

CAS RN 446-24-2

NH2HNOF

CAS RN 446-24-2

IUPAC Name: 2-fluorobenzohydrazide
Synonyms: 2-fluorobenzohydrazide
Molecular Weight (g/mol): 154.14
Molecular Formula: C7H7FN2O
InChi Key: YJCCKQQVXNNAAR-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC=C1F
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)