missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzoic hydrazide, 97%, Thermo Scientific™

Product Code. 11985671 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11985671

Brand: Thermo Scientific Acros 443370100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 446-24-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 70.0°C to 72.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H7FN2O
MDL Number MFCD00025112
Quantity 10 g
Synonym 2-fluorobenzhydrazide, 2-fluorobenzoic hydrazide, o-fluorobenzhydrazide, 2-fluorobenzoic acid hydrazide, 2-fluorobenzoylhydrazine, 2-fluorobenzenecarbohydrazide, pubchem3481, 2-fluoro-benzohydrazide, acmc-209jyw, 2-fluoranylbenzohydrazide
InChI Key YJCCKQQVXNNAAR-UHFFFAOYSA-N
SMILES NNC(=O)C1=CC=CC=C1F
IUPAC Name 2-fluorobenzohydrazide
Molecular Weight (g/mol) 154.14
PubChem CID 136288
Formula Weight 154.14
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Fluorobenzoic hydrazide
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluorobenzoic hydrazide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.