accessibility menu, dialog, popup

UserName

H3CONFFF

CAS RN 447-93-8

IUPAC Name: 3-methoxy-4-(trifluoromethyl)benzonitrile
Synonyms: 3-methoxy-4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 201.15
Molecular Formula: C9H6F3NO
InChi Key: QASUCJMIPAXDOB-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(=C1)C#N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results