CAS RN 448184-56-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
S-Desethylpiperidino S-[2-Carboxyl-2-oxo-ethyl-(2-aminoethyl)amino] Vardenafil, TRC
CAS: 448184-56-3 Molecular Formula: C21H26N6O7S Molecular Weight (g/mol): 506.53 Synonym: 2-[[2-[[[3-(1,4-Dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]amino]ethyl]amino]-2-oxoacetic Acid,[[2-[[[3-(1,4-Dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]amino]ethyl]amino]oxoacetic Acid,Vardenafil Oxoacetic Acid Impurity SMILES: O=S(NCCNC(C(O)=O)=O)(C1=CC=C(OCC)C(C2=NN3C(C(N2)=O)=C(C)N=C3CCC)=C1)=O