missing translation for 'onlineSavingsMsg'
Learn More

S-Desethylpiperidino S-[2-Carboxyl-2-oxo-ethyl-(2-aminoethyl)amino] Vardenafil, TRC

Product Code. 30136275
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136275 25 mg 25mg
30135256 10 mg 10mg
17900415 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136275 Supplier Toronto Research Chemicals Supplier No. TRCD29097025MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 448184-56-3
Molecular Formula C21H26N6O7S
Molecular Weight (g/mol) 506.53
Synonym 2-[[2-[[[3-(1,4-Dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]amino]ethyl]amino]-2-oxoacetic Acid, [[2-[[[3-(1,4-Dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]amino]ethyl]amino]oxoacetic Acid, Vardenafil Oxoacetic Acid Impurity
SMILES O=S(NCCNC(C(O)=O)=O)(C1=CC=C(OCC)C(C2=NN3C(C(N2)=O)=C(C)N=C3CCC)=C1)=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H26N6O7S/c1-4-6-16-24-12(3)17-19(28)25-18(26-27(16)17)14-11-13(7-8-15(14)34-5-2)35(32,33)23-10-9-22-20(29)21(30)31/h7-8,11,23H,4-6,9-10H2,1-3H3,(H,22,29)(H,30,31)(H,25,26,28)
Formula Weight 506.1584
Chemical Name or Material S-Desethylpiperidino S-[2-Carboxyl-2-oxo-ethyl-(2-aminoethyl)amino] Vardenafil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.