CAS RN 448950-89-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
7-Chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine, TRC
CAS: 448950-89-8 Molecular Formula: C17 H13 Cl N4 Molecular Weight (g/mol): 308.76 Synonym: Alprazolam Imp. G (EP),7-Chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine IUPAC Name: 7-chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine SMILES: Cc1nnc2c(N)c(c3ccccc3)c4cc(Cl)ccc4n12
2
7-Chloro-1-methyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-4-amine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.