accessibility menu, dialog, popup

UserName

CAS RN 451-02-5

H3COOF

CAS RN 451-02-5

IUPAC Name: ethyl 3-fluorobenzoate
Synonyms: ethyl 3-fluorobenzoate
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: SMMIKBXLEWTSJD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results