accessibility menu, dialog, popup

UserName

CH3FHO

CAS RN 452-85-7

IUPAC Name: 5-fluoro-2-methylphenol
Synonyms: 5-fluoro-2-methylphenol
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: CKJOSIHYVLHIER-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results