missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylphenol, 98%, Thermo Scientific™

Product Code. 11411637
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411637 5 g 5g
11401637 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411637 Supplier Thermo Scientific Alfa Aesar Supplier No. B25606.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-85-7
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00673009
InChI Key CKJOSIHYVLHIER-UHFFFAOYSA-N
Synonym 4-fluoro-2-hydroxytoluene, 5-fluoro-o-cresol, 5-fluoro-2-methyl-phenol, pubchem1502, 2-methyl-5-fluorophenol, acmc-1afs5, 5-fluoranyl-2-methyl-phenol, ksc494a2f
PubChem CID 2774616
IUPAC Name 5-fluoro-2-methylphenol
SMILES CC1=C(C=C(C=C1)F)O

Specifications

Density 1.157
Boiling Point 66°C (4 mmHg)
Odor Phenol-like
Refractive Index 1.514
Quantity 5 g
UN Number UN3265
Beilstein 3234863
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-methylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.