accessibility menu, dialog, popup

UserName

CAS RN 454-92-2

IUPAC Name: 3-(trifluoromethyl)benzoic acid
Synonyms: 3-trifluoromethyl-benzoic acid 3-tfmba α,α,α-trifluoro-m-toluic acid M-trifluoromethylbenzoic acid
Molecular Weight (g/mol): 190.12
Molecular Formula: C8H5F3O2
InChi Key: FQXQBFUUVCDIRK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C(F)(F)F
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)