accessibility menu, dialog, popup

UserName

FFFI

CAS RN 455-13-0

IUPAC Name: 1-iodo-4-(trifluoromethyl)benzene
Synonyms: 1-iodo-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 272.01
Molecular Formula: C7H4F3I
InChi Key: SKGRFPGOGCHDPC-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)