missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzotrifluoride, 98%

Product Code. 11494833
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11494833 1 g 1g
11404843 5 g 5g
11414843 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11494833

Brand: Thermo Scientific Alfa Aesar A12189.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 455-13-0 | C7H4F3I | 272.009 g/mol

4-Iodobenzotrifluoride may be employed as substrate with electron-deficient aromatic ring, during Mizoroki-Heck reaction with acrylic acid, to afford 4-trifluoromethylcinnnamic acid. It undergoes aminocarbonylation in DMF using phosphoryl chloride to give N,N-dimethyl-(4-trifluoromethyl)benzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzotrifluoride may be employed as substrate with electron-deficient aromatic ring, during Mizoroki-Heck reaction with acrylic acid, to afford 4-trifluoromethylcinnnamic acid. It undergoes aminocarbonylation in DMF using phosphoryl chloride to give N,N-dimethyl-(4-trifluoromethyl)benzamide.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents. Store in dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-13-0
Molecular Formula C7H4F3I
Molecular Weight (g/mol) 272.009
MDL Number MFCD00039398
InChI Key SKGRFPGOGCHDPC-UHFFFAOYSA-N
Synonym 4-iodobenzotrifluoride, 1-iodo-4-trifluoromethyl benzene, p-iodobenzotrifluoride, benzene, 1-iodo-4-trifluoromethyl, 4-trifluoromethyl iodobenzene, 4-iodo-alpha,alpha,alpha-trifluorotoluene, alpha,alpha,alpha-trifluoro-4-iodotoluene, 4-iodobenztrifluoride, 4-iodobenzo trifluoride, 4-iodo-1-trifluoromethylbenzene
PubChem CID 67993
IUPAC Name 1-iodo-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)I

Specifications

Melting Point -8°C
Density 1.851
Boiling Point 68°C to 70°C (15 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.518
Quantity 1 g
Beilstein 1944013
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 272.01
Percent Purity 98%
Chemical Name or Material 4-Iodobenzotrifluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.