accessibility menu, dialog, popup

UserName

CAS RN 455-19-6

IUPAC Name: 4-(trifluoromethyl)benzaldehyde
Synonyms: 4-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 174.12
Molecular Formula: C8H5F3O
InChi Key: BEOBZEOPTQQELP-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=O)C=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)