missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzaldehyde, 97%

Product Code. 11452338
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452338 25 g 25g
11442338 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452338 Supplier Thermo Scientific Alfa Aesar Supplier No. A15276.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 455-19-6 | C8H5F3O | 174.122 g/mol

4-(Trifluoromethyl)benzaldehyde is useful reagent in kinetic studies of the asymmetric synthesis of alcohols and of the Wittig reaction. It is also used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzaldehyde is useful reagent in kinetic studies of the asymmetric synthesis of alcohols and of the Wittig reaction. It is also used as Pharmaceutical intermediates.

Solubility
Soluble in water. 1.5 g/L at 20°C

Notes
Air Sensitive. Store away from air and oxidizing agents. Store under an inert atmosphere. Incompatible with oxidizing agents, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-19-6
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.122
MDL Number MFCD00006952
InChI Key BEOBZEOPTQQELP-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl-benzaldehyde
PubChem CID 67996
IUPAC Name 4-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=CC=C1C=O)C(F)(F)F

Specifications

Melting Point 1°C to 2°C
Density 1.275
Boiling Point 80°C to 81°C (25 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.463
Quantity 25 g
Beilstein 1101680
Sensitivity Air Sensitive
Solubility Information Soluble in water. 1.5g/L at 20°C
Formula Weight 174.12
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.