accessibility menu, dialog, popup

UserName

H3COF

CAS RN 456-49-5

IUPAC Name: 1-fluoro-3-methoxybenzene
Synonyms: M-fluoroanisole
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: MFJNOXOAIFNSBX-UHFFFAOYSA-N
SMILES: COC1=CC=CC(F)=C1
Boiling Point 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)