missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoroanisole, 98%, Thermo Scientific™

Product Code. 10731641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10731641 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10731641 Supplier Thermo Scientific Acros Supplier No. 171890250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 456-49-5
CAS Min % 98.5
CAS Max % 100.0
Melting Point -35.0°C
Color Colorless to Straw-Yellow
Density 1.1040g/mL
Boiling Point 158.0°C (743.0 mmHg)
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7FO
Linear Formula FC6H4OCH3
Refractive Index 1.4876 to 1.4896
MDL Number MFCD00000335
Quantity 25 g
Specific Gravity 1.104
Synonym 3-fluoroanisole, m-fluoroanisole, benzene, 1-fluoro-3-methoxy, 1-fluoro-3-methoxy-benzene, 2-fluoro-4-methoxybenzene, fr co1 wln, 3-fluoroanisol, 3-fluoro-anisole, 3-fluoro anisole, anisole, m-fluoro
InChI Key MFJNOXOAIFNSBX-UHFFFAOYSA-N
SMILES COC1=CC=CC(F)=C1
IUPAC Name 1-fluoro-3-methoxybenzene
Molecular Weight (g/mol) 126.13
PubChem CID 9975
Formula Weight 126.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Fluoroanisole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluoroanisole
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.