accessibility menu, dialog, popup

UserName

CAS RN 4654-39-1

OHBr

CAS RN 4654-39-1

IUPAC Name: 2-(4-bromophenyl)ethan-1-ol
Synonyms: 2-(4-bromophenyl)ethanol
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: PMOSJSPFNDUAFY-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(Br)C=C1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)