missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenethyl alcohol, 99%

Product Code. 10184430 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
10g; Glass bottle
This item is not returnable. View return policy

Product Code. 10184430

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4654-39-1 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4654-39-1
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4300g/mL
Boiling Point 96°C to 99°C (0.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H9BrO
Linear Formula BrC6H4CH2CH2OH
Refractive Index 1.5720 to 1.574
MDL Number MFCD00002897
Quantity 10 g
Beilstein 06, III, 1713
Specific Gravity 1.43
Synonym 2-4-bromophenyl ethanol, 4-bromophenethyl alcohol, p-bromophenethyl alcohol, benzeneethanol, 4-bromo, 2-4-bromophenyl ethan-1-ol, 2-4-bromo-phenyl-ethanol, 2-4-bromophenyl ethyl alcohol, 2-p-bromophenyl ethanol, phenethyl alcohol, p-bromo, 4-bromobenzene ethanol
InChI Key PMOSJSPFNDUAFY-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CCO)Br
IUPAC Name 2-(4-bromophenyl)ethanol
Molecular Weight (g/mol) 201.06
PubChem CID 72851
Formula Weight 201.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromophenethyl alcohol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromophenethyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.