accessibility menu, dialog, popup

UserName

CH3H3C(S)OH(S)H3C(R)(R)H3C(R)O(R)(S)CH3(S)H3C(S)CH3HOO

CAS RN 471-53-4

IUPAC Name: (2S,4aS,6aS,6bR,8aR,10S,12aS,12bR,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-2-carboxylic acid
Synonyms: jintan acid, uralenic glyciram glycyrrhetinic acid (product) α-glycyrrhetinic acid 18β-glycyrrhetinic acid acid, rhetinic acid, glycyrrhetinic glycyram glycyrrhetin
Molecular Weight (g/mol): 470.69
Molecular Formula: C30H46O4
InChi Key: MPDGHEJMBKOTSU-YKLVYJNSSA-N
SMILES: CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results