accessibility menu, dialog, popup

UserName

CAS RN 4746-97-8

OOO

CAS RN 4746-97-8

IUPAC Name: 1,4-dioxaspiro[4.5]decan-8-one
Synonyms: 1,4-dioxaspiro[4.5]decan-8-one
Molecular Weight (g/mol): 156.18
Molecular Formula: C8H12O3
InChi Key: VKRKCBWIVLSRBJ-UHFFFAOYSA-N
SMILES: O=C1CCC2(CC1)OCCO2
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (2)
  • (2)
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (5)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)