accessibility menu, dialog, popup

UserName

CAS RN 480-18-2

HO(R)(R)OOHOHOHOOH

CAS RN 480-18-2

IUPAC Name: (2R,3R)-2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: (+)-taxifolin taxifolin dihydroquercetin (+)-dihydroquercetin (2R,3R)-dihydroquercetin trans-dihydroquercetin dihydro-quercetin distylin (2s,3s)-trans-dihydroquercetin taxifoliol
Molecular Weight (g/mol): 304.25
Molecular Formula: C15H12O7
InChi Key: CXQWRCVTCMQVQX-LSDHHAIUSA-N
SMILES: O[C@@H]1[C@H](OC2=CC(O)=CC(O)=C2C1=O)C1=CC=C(O)C(O)=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results