missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals (+)-trans-Taxifolin
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 500 mg |
| Melting Point | 216.0°C to 220.0°C |
| Formula Weight | 304.25 |
| InChI Key | CXQWRCVTCMQVQX-LSDHHAIUSA-N |
| SMILES | O[C@@H]1[C@H](OC2=CC(O)=CC(O)=C2C1=O)C1=CC=C(O)C(O)=C1 |
| IUPAC Name | (2R,3R)-2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-3,4-dihydro-2H-1-benzopyran-4-one |
| Molecular Weight (g/mol) | 304.25 |
| Chemical Name or Material | (+)-trans-Taxifolin |
Safety and Handling
Product Identifier
- (+)-trans-Taxifolin
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : (+)-trans-Taxifolin
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction