accessibility menu, dialog, popup

UserName

CAS RN 48208-26-0

ONOHNOHO

CAS RN 48208-26-0

IUPAC Name: 2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-3-(1H-indol-3-yl)propanoic acid
Synonyms: 2-(1,3-dioxoisoindol-2-yl)-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol): 334.33
Molecular Formula: C19H14N2O4
InChi Key: HPTXLHAHLXOAKV-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CNC2=CC=CC=C12)N1C(=O)C2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results