CAS RN 4840-81-7
CAS RN 4840-81-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-Methyl-2-pentanone-1,1,1,3,3-d5, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
4-Methyl-2-pentanone-d5, TRC
CAS: 4840-81-7 Molecular Formula: C6 2H5 H7 O Molecular Weight (g/mol): 105.19 Synonym: 4-Methyl-2-pentanone D5 (1,1,1,3,3-D5),2-Pentanone-1,1,1,3,3-d5, 4-methyl- (7CI, 8CI, 9CI) IUPAC Name: 1,1,1,3,3-pentadeuterio-4-methylpentan-2-one SMILES: [2H]C([2H])([2H])C(=O)C([2H])([2H])C(C)C