CAS RN 500-92-5
CAS RN 500-92-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Proguanil, MedChemExpress
MedChemExpress Proguanil, an antimalarial prodrug, is metabolized to the active metabolite Cycloguanil (HY-12784). Proguanil is a dihydrofolate reductase (DHFR) inhibitor.