accessibility menu, dialog, popup

UserName

ONHOHN

CAS RN 504-07-4

IUPAC Name: 1,3-diazinane-2,4-dione
Synonyms: 5,6-dihydrouracil 3,4-dihydrouracil
Molecular Weight (g/mol): 114.1
Molecular Formula: C4H6N2O2
InChi Key: OIVLITBTBDPEFK-UHFFFAOYSA-N
SMILES: O=C1CCNC(=O)N1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results