missing translation for 'onlineSavingsMsg'
Learn More

Dihydro Uracil, TRC

Product Code. 30012643
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012643 1 g 1g
30008988 100 mg 100mg
30012092 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012643 Supplier Toronto Research Chemicals Supplier No. TRCD4499901G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-07-4
Molecular Formula C4 H6 N2 O2
Molecular Weight (g/mol) 114.1
Synonym 2,4(1H,3H)-Pyrimidinedione, dihydro- (9CI, ACI), Dihydro-2,4(1H,3H)-pyrimidinedione (ACI), Hydrouracil (6CI, 8CI), 1,3-Diazinane-2,4-dione, 5,6-Dihydro-2,4(1H,3H)-pyrimidinedione, 5,6-Dihydro-2,4-dihydroxypyrimidine, 5,6-Dihydrouracil, Dihydrouracil, Hexahydropyrimidine-2,4-dione, NSC 11867
IUPAC Name 1,3-diazinane-2,4-dione
SMILES O=C1CCNC(=O)N1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H6N2O2/c7-3-1-2-5-4(8)6-3/h1-2H2,(H2,5,6,7,8)
Formula Weight 114.0429
Percent Purity >95%
Chemical Name or Material Dihydro Uracil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.