accessibility menu, dialog, popup

UserName

CAS RN 5074-71-5

IUPAC Name: bis(2,3,4,5,6-pentafluorophenyl)(phenyl)phosphane
Synonyms: bis(2,3,4,5,6-pentafluorophenyl)(phenyl)phosphane
Molecular Weight (g/mol): 442.2
Molecular Formula: C18H5F10P
InChi Key: OYNXPGGNQMSMTR-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
Molecular Weight (g/mol) 
  • (1)
  • (8)
Solvent or Matrix 
  • (1)
  • (1)
  • (2)
  • (1)
Concentration 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results