accessibility menu, dialog, popup

UserName

POOHBr

CAS RN 50889-29-7

IUPAC Name: (5-carboxypentyl)triphenylphosphanium bromide
Synonyms: (5-carboxypentyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 457.35
Molecular Formula: C24H26BrO2P
InChi Key: JUWYRPZTZSWLCY-UHFFFAOYSA-N
SMILES: [Br-].OC(=O)CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)