missing translation for 'onlineSavingsMsg'
Learn More

(5-Carboxypentyl)(triphenyl)phosphonium bromide, 97%, Thermo Scientific™

Product Code. 11316885
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11316885 25 g Amber Glass Bottle 25g
10463173 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11316885 Supplier Thermo Scientific Maybridge Supplier No. KM08684EB
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 50889-29-7
Molecular Formula C24H26BrO2P
Molecular Weight (g/mol) 457.35
MDL Number MFCD00055556
InChI Key JUWYRPZTZSWLCY-UHFFFAOYSA-N
Synonym 5-carboxypentyl triphenylphosphonium bromide, 5-carboxypentyl triphenyl phosphonium bromide, 5-carboxypentyl triphenylphosphanium bromide, 5-carboxypentyltriphenylphosphonium bromide, phosphonium, 5-carboxypentyl triphenyl-, bromide, acmc-20apmi, pxjvnuuwrjumaq-uhfffaoysa-m, 5-carboxyamyl triphenylphosphonium bromide
PubChem CID 2779280
IUPAC Name 5-carboxypentyl(triphenyl)phosphanium;bromide
SMILES [Br-].OC(=O)CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 457.35
Percent Purity 97%
Chemical Name or Material (5-Carboxypentyl)(triphenyl)phosphonium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.