accessibility menu, dialog, popup

UserName

HONH2

CAS RN 51135-96-7

IUPAC Name: 4-benzyl-4-hydroxypiperidin-1-ium
Synonyms: 4-benzyl-4-hydroxypiperidin-1-ium
Molecular Weight (g/mol): 192.28
Molecular Formula: C12H18NO
InChi Key: KJZBZOFESQSBCV-UHFFFAOYSA-O
SMILES: OC1(CC2=CC=CC=C2)CC[NH2+]CC1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)