missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-4-hydroxypiperidine, 97%

Product Code. 11497587
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497587 25 g 25g
11477587 1 g 1g
11487587 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497587 Supplier Thermo Scientific Alfa Aesar Supplier No. A15664.14
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51135-96-7 | C12H17NO | 191.274 g/mol

It is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51135-96-7
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00023147
InChI Key KJZBZOFESQSBCV-UHFFFAOYSA-N
Synonym 4-benzyl-4-hydroxypiperidine, 4-piperidinol, 4-benzyl, 4-benzyl-4-piperidinol, 4-piperidinol, 4-phenylmethyl, acmc-20amoy, pubchem16262, 4benzyl-piperidin-4-ol, maybridge3_002978, 4-benzyl-piperidin-4-ol, 4-benzyl-4-piperidinol #
PubChem CID 98152
IUPAC Name 4-benzylpiperidin-4-ol
SMILES C1CNCCC1(CC2=CC=CC=C2)O

Specifications

Melting Point ∼80°C
Quantity 25 g
Beilstein 1451803
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 191.27
Percent Purity 97%
Chemical Name or Material 4-Benzyl-4-hydroxypiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.