missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-4-hydroxypiperidine, 95%, Thermo Scientific™

Product Code. 10050710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10050710 1 g Glass bottle 1g
10549620 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10050710 Supplier Thermo Scientific Acros Supplier No. 105990010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51135-96-7
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.27
MDL Number MFCD00023147
InChI Key KJZBZOFESQSBCV-UHFFFAOYSA-N
Synonym 4-benzyl-4-hydroxypiperidine, 4-piperidinol, 4-benzyl, 4-benzyl-4-piperidinol, 4-piperidinol, 4-phenylmethyl, acmc-20amoy, pubchem16262, 4benzyl-piperidin-4-ol, maybridge3_002978, 4-benzyl-piperidin-4-ol, 4-benzyl-4-piperidinol #
PubChem CID 98152
IUPAC Name 4-benzylpiperidin-4-ol
SMILES C1CNCCC1(CC2=CC=CC=C2)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 80.0°C to 85.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 191.27
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyl-4-hydroxypiperidine, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzyl-4-hydroxypiperidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.