accessibility menu, dialog, popup

UserName

CH3OOOHOOH3COOH3COHOOH3COOCH3OOHOCH3OOH3COOHOCH3OOCH3OHOOH3COOCH3OHOOH3COOCH3OHOOH3C

CAS RN 51166-71-3

IUPAC Name: 37,39,41,43,45,47,49-heptamethoxy-5,10,15,20,25,30,35-heptakis(methoxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,38,40,42,44,46,48-heptol
Synonyms: methyl cyclodextrin
Molecular Weight (g/mol): 1331.37
Molecular Formula: C56H98O35
InChi Key: QGKBSGBYSPTPKJ-UHFFFAOYNA-N
SMILES: COCC1OC2OC3C(COC)OC(OC4C(COC)OC(OC5C(COC)OC(OC6C(COC)OC(OC7C(COC)OC(OC8C(COC)OC(OC1C(O)C2OC)C(OC)C8O)C(OC)C7O)C(OC)C6O)C(OC)C5O)C(OC)C4O)C(OC)C3O
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)