missing translation for 'onlineSavingsMsg'
Learn More

Di-O-methyl-beta-cyclodextrin, TRC

Product Code. 30136796
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136796 5 g 5g
30133213 10 g 10g
30005773 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136796 Supplier Toronto Research Chemicals Supplier No. TRCD4708705G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51166-71-3
Molecular Formula C56 H98 O35
Molecular Weight (g/mol) 1331.36
Synonym 2A,2B,2C,2D,2E,2F,2G,6A,6B,6C,6D,6E,6F,6G-Tetradeca-O-methyl-β-cyclodextrin, 2,6-Dimethyl-β-cyclodextrin, Dimeb, Hepta-(2,6-di-oxy-methyl)-β-cyclodextrin, Heptakis(2,6-di-O-methyl)-β-cyclodextrin, Tetradeca-O-methyl-β-cyclodextrin, Tetradecakis-2,6-O-methylcycloheptaamylose, (DMβCD);
SMILES COC[C@H]1O[C@@H]2O[C@@H]3[C@@H](COC)O[C@H](O[C@@H]4[C@@H](COC)O[C@H](O[C@@H]5[C@@H](COC)O[C@H](O[C@@H]6[C@@H](COC)O[C@H](O[C@@H]7[C@@H](COC)O[C@H](O[C@@H]8[C@@H](COC)O[C@H](O[C@H]1[C@H](O)[C@H]2OC)[C@H](OC)[C@H]8O)[C@H](OC)[C@H]7O)[C@H](OC)[C@H]6O)[C@H](OC)[C@H]5O)[C@H](OC)[C@H]4O)[C@H](OC)[C@H]3O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C56H98O35/c1-64-15-22-36-29(57)43(71-8)50(78-22)86-37-23(16-65-2)80-52(45(73-10)30(37)58)88-39-25(18-67-4)82-54(47(75-12)32(39)60)90-41-27(20-69-6)84-56(49(77-14)34(41)62)91-42-28(21-70-7)83-55(48(76-13)35(42)63)89-40-26(19-68-5)81-53(46(74-11)33(40)61)87-38-24(17-66-3)79-51(85-36)44(72-9)31(38)59/h22-63H,15-21H2,1-14H3/t22-,23-,24-,25-,26-,27-,28-,29+,30+,31+,32+,33+,34+,35+,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,55-,56-/m1/s1
Formula Weight 1330.5889
Chemical Name or Material Di-O-methyl-beta-cyclodextrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.