missing translation for 'onlineSavingsMsg'
Learn More

Heptakis(2,6-di-o-methyl)-β-cyclodextrin, MP Biomedicals™

Product Code. 11463802 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463802 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11463802 Supplier MP Biomedicals Supplier No. 0215732005
  • ONLINE EXCLUSIVE PRICE
    403.00 €
    382.20 € / 5g
    Save 20.80 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Heptakis(2,6-di-O-methyl)-β-cyclodextrin is a cholesterol sequestering agent

Chemical Identifiers

CAS 51166-71-3
Molecular Formula C56H98O35
Molecular Weight (g/mol) 1331.365
MDL Number MFCD00011616
InChI Key QGKBSGBYSPTPKJ-UZMKXNTCSA-N
Synonym 2,6-di-o-methyl-beta-cyclodextrin, unii-5fjp73e35o, dimethyl-beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis-2,6-di-o-methyl-beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, dm-beta-cyd, dimethyl, a-cyclodextrin, 2,6-dme-beta-cyclodextrin, 2,6-di-o-methyl-beta-cd
PubChem CID 10171019
SMILES COCC1C2C(C(C(O1)OC3C(OC(C(C3O)OC)OC4C(OC(C(C4O)OC)OC5C(OC(C(C5O)OC)OC6C(OC(C(C6O)OC)OC7C(OC(C(C7O)OC)OC8C(OC(O2)C(C8O)OC)COC)COC)COC)COC)COC)COC)OC)O

Specifications

Color White
Quantity 5 g
Formula Weight 1331.4
Physical Form Powder
Chemical Name or Material Heptakis(2, 6-di-o-methyl)-β-cyclodextrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.