CAS RN 514222-44-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1,1'-(1,3-Phenylene)di-1H-1,2,4-triazole, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
1,1'-(1,3-Phenylene)bis-1H-1,2,4-triazole, TRC
CAS: 514222-44-7 Molecular Formula: C10 H8 N6 Molecular Weight (g/mol): 212.21 Synonym: 1,1'-(1,3-Phenylene)di-1H-1,2,4-triazole,Fluconazole Imp. C (EP) IUPAC Name: 1-[3-(1,2,4-triazol-1-yl)phenyl]-1,2,4-triazole SMILES: c1cc(cc(c1)n2cncn2)n3cncn3