accessibility menu, dialog, popup

UserName

CH3NOO(E/Z)

CAS RN 5153-68-4

IUPAC Name: 1-methyl-4-(2-nitroethenyl)benzene
Synonyms: 4-methyl-B-nitrostyrene
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: JSPNBERPFLONRX-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C[N+]([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results