accessibility menu, dialog, popup

UserName

H3COOCH3H3C

CAS RN 53012-41-2

IUPAC Name: 1-(2,6-dimethylphenoxy)propan-2-one
Synonyms: 1-(2,6-dimethylphenoxy)propan-2-one
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: XDJULAUHYAJQBU-UHFFFAOYSA-N
SMILES: CC(=O)COC1=C(C)C=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results