accessibility menu, dialog, popup

UserName

CAS RN 5315-79-7

IUPAC Name: pyren-1-ol
Synonyms: 1-hydroxypyrene
Molecular Weight (g/mol): 218.26
Molecular Formula: C16H10O
InChi Key: BIJNHUAPTJVVNQ-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)