missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxypyrene, TRC

Product Code. 30133659
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133659 250 mg 250mg
17904935 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133659 Supplier Toronto Research Chemicals Supplier No. TRCH952700250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5315-79-7
Molecular Formula C16 H10 O
Molecular Weight (g/mol) 218.25
Synonym 1-Pyrenol, 1-Hydroxypyrene, 3-Hydroxypyrene, 3-Pyrenol, NSC 30968
IUPAC Name pyren-1-ol
SMILES Oc1ccc2ccc3cccc4ccc1c2c34

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C16H10O/c17-14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9,17H
Formula Weight 218.0732
Percent Purity >95%
Chemical Name or Material 1-Hydroxypyrene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.