accessibility menu, dialog, popup

UserName

NH2IOHO

CAS RN 5326-47-6

IUPAC Name: 2-amino-5-iodobenzoic acid
Synonyms: 2-amino-5-iodobenzoic acid
Molecular Weight (g/mol): 263.03
Molecular Formula: C7H6INO2
InChi Key: GOLGILSVWFKZRQ-UHFFFAOYSA-N
SMILES: NC1=CC=C(I)C=C1C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results