missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-iodobenzoic acid, 98%

Product Code. 11448687
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448687 25 g 25g
11438687 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448687 Supplier Thermo Scientific Alfa Aesar Supplier No. A19838.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5326-47-6 | C7H6INO2 | 263.034 g/mol

2-Amino-5-iodobenzoic acid is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as pharmaceutical intermediates, polarizing films for Liquid Crystal Display (LCD) chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-iodobenzoic acid is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible with light and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5326-47-6
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.034
MDL Number MFCD00007849
InChI Key GOLGILSVWFKZRQ-UHFFFAOYSA-N
Synonym 5-iodoanthranilic acid, benzoic acid, 2-amino-5-iodo, 5-iodoanthranil acid, anthranilic acid, 5-iodo, 2-amino-5-iodo-benzoic acid, pubchem2543, 5-iodo anthranilic acid, acmc-1ays8, 2-amino-5-iodobenzoicacid, 5-iodo-2-aminobenzoic acid
PubChem CID 72911
IUPAC Name 2-amino-5-iodobenzoic acid
SMILES C1=CC(=C(C=C1I)C(=O)O)N

Specifications

Melting Point ∼220°C (decomposition)
Quantity 25 g
Beilstein 639029
Sensitivity Light sensitive
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 263.03
Percent Purity 98%
Chemical Name or Material 2-Amino-5-iodobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.