accessibility menu, dialog, popup

UserName

CH3ONHCl

CAS RN 533-17-5

IUPAC Name: N-(2-chlorophenyl)acetamide
Synonyms: 2'-chloroacetanilide
Molecular Weight (g/mol): 169.61
Molecular Formula: C8H8ClNO
InChi Key: KNVQTRVKSOEHPU-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results