accessibility menu, dialog, popup

UserName

H3CNCH3SOO

CAS RN 5339-59-3

IUPAC Name: N,N-dibutylbenzenesulfonamide
Synonyms: N,N-dibutylbenzenesulfonamide
Molecular Weight (g/mol): 269.4
Molecular Formula: C14H23NO2S
InChi Key: YKXINVWHNNYQMA-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)S(=O)(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results