accessibility menu, dialog, popup

UserName

H3COBrOCH3NOO

CAS RN 53413-67-5

IUPAC Name: 1-(bromomethyl)-4,5-dimethoxy-2-nitrobenzene
Synonyms: 1-(bromomethyl)-4,5-dimethoxy-2-nitrobenzene
Molecular Weight (g/mol): 276.09
Molecular Formula: C9H10BrNO4
InChi Key: UEKFEYNZISYRRH-UHFFFAOYSA-N
SMILES: COC1=CC(CBr)=C(C=C1OC)[N+]([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results