missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzyl bromide, 97%

Product Code. 11944721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11944721 5 g 5g
11386831 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11944721 Supplier Thermo Scientific Acros Supplier No. 440770050
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 53413-67-5 | C9H10BrNO4 | 276.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53413-67-5
Molecular Formula C9H10BrNO4
Molecular Weight (g/mol) 276.09
InChI Key UEKFEYNZISYRRH-UHFFFAOYSA-N
Synonym 1-bromomethyl-4,5-dimethoxy-2-nitrobenzene, 4,5-dimethoxy-2-nitrobenzyl bromide, 1-bromomethyl-2-nitro-4,5-dimethoxybenzene, bmndmb, benzene, 1-bromomethyl-4,5-dimethoxy-2-nitro, 6-nitroveratryl bromide, acmc-1ao6p, 4,5-dimenoxy-2-nitrobenzyl bromide, 2-nitro-4,5-dimethoxybenzyl bromide, 1-bromomethyl-4,5-dimethoxynitrobenzene
PubChem CID 3016812
IUPAC Name 1-(bromomethyl)-4,5-dimethoxy-2-nitrobenzene
SMILES COC1=C(C=C(C(=C1)CBr)[N+](=O)[O-])OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 130°C to 132°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 276.09
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 4, 5-Dimethoxy-2-nitrobenzyl bromide
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • 4,5-Dimethoxy-2-nitrobenzyl bromide
Avvertenza
  • Danger
Categoria di pericolo
  • Skin corrosion/irritation Category 1 B
Indicazioni di pericolo
  • H314-Causes severe skin burns and eye damage.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.